Choline hydrogen tartrate 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 2-Hydroxy-N,N,N-trimethylethan-1-aminium (2R,3R)-3-carboxy-2,3-dihydroxypropanoate, Choline acid tartrate, CBT, 2-(Hydroxyethyl)trimethylammonium bitartrate, Choline bitartrate
CAS Number:87-67-2
:
Formula:C₉H₁₉NO₇
MDL Number:MFCD00036332
Molecular Weight:253.25 g/mol
Melting Pt:151…153 °C
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Water:≤0.5 %
- Titration with HClO4:≥96.0 %
Specification Test Results
Frequently Bought Together