Sodium phosphite dibasic pentahydrate 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Etidronate disodium Related Compound A, United States Pharmacopeia (USP) Reference Standard, METAPHOSPHORIC ACID SODIUM SALT
CAS Number:13517-23-2
:
Formula:H₁₁Na₂O₈P
MDL Number:MFCD00150354
Molecular Weight:216.033 g/mol
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Transparent white to white Crystals and or bars
- Titration with HCl:≥97.5 %
- Chloride (Cl):≤50 ppm
- Sulfate (SO4):≤50 ppm
- Iron (Fe):≤10 ppm
- Heavy metals (as Pb):≤10 ppm
Specification Test Results
Frequently Bought Together