Order Entry
Austria
ContactUsLinkComponent
3-Hydroxy-4-nitrobenzaldehyde 97%
3-Hydroxy-4-nitrobenzaldehyde 97%
Certificates
undefined
3-Hydroxy-4-nitrobenzaldehyde 97%

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    Yellow to brown Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    130 to 135 °C
  • GC:
    ≥96.0 %

Specification Test Results

Related Information

Frequently Bought Together