2-Chloroquinoxaline 97%
:1448-87-9
:
:C₈H₅ClN₂
:MFCD00043907
:100 °C (1.4 mmHg)
:164.5917 g/mol
:48…50 °C
:GHS07
:Ambient
- Environmentally Preferable:
- Appearance:White to grey to brown Crystalline powder or solid
- Infrared spectrum:Conforms
- Melting point:47 to 51 °C
- HPLC:≥96.0 %
Frequently Bought Together