5-Aminoquinoline
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Quinolin-5-amine, Quinolin-5-ylamine
CAS Number:611-34-7
:
Formula:C₉H₈N₂
MDL Number:MFCD00006797
Boiling Pt:310
Molecular Weight:144.18 g/mol
Melting Pt:106…109 °C
:GHS07
Storage Temperature:Refrigerator
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Yellow to brown Crystalline powder
- Infrared spectrum:Conforms
- Melting point:106 to 112 °C
- GC:≥96.0 %
Specification Test Results
Frequently Bought Together