2-Methyl-1H-indole-3-carbaldehyde 97+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 2-Methylindole-3-carboxaldehyde, 3-Formyl-2-methylindole
CAS Number:5416-80-8
:
Formula:C₁₀H₉NO
MDL Number:MFCD00012077
Molecular Weight:159.1846 g/mol
Melting Pt:204…205 °C
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Brown Powder
- Infrared spectrum:Conforms
- Melting point:199 to 202 °C
- HPLC:≥97.0 %
Specification Test Results
Frequently Bought Together