Ruthenium red tetrahydrate, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: AMMONIATED RUTHENIUM OXYCHLORIDE, RUTHENIUM CHLORIDE OXIDE, AMMONIATE, Ruthenium(6+),tetradecaamminedi-m-oxotri-, hexachloride, tetrahydrate(9CI)
CAS Number:12790-48-6
:
Formula:Cl₆H₄₂N₁₄O₂Ru₃·4H₂O
MDL Number:MFCD00167176
Molecular Weight:858.42 g/mol
:GHS07
Storage Temperature:Refrigerator
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Dark red-purple Crystalline powder
- Assay:34.6 to 36.1 % (Ru)
- UV Lambda max:530 to 536 nm
- Molar extinction coefficient:≥8000 l/(mole.cm) (lambda max., 0.02 g/l H2O)
Specification Test Results
Frequently Bought Together