Racks for PCR tubes
Supplier: NEOLAB
Certificates
About this item
Racks, PP, bright colors (orange, yellow, green, pink, blue). Especially designed for 0.2 ml PCR tubes in strips.
- Accommodates strips of 8 or 12 tubes
- Holds 96 PCR-tubes (8 x 12 positions)
- Alphanumerical index
- Stackable
- Complete with transparent cover
Frequently Bought Together