Marfey's reagent (FDAA or 1-fluoro-2-4-dinitrophenyl-5-L-alanine amide), Pierce™
Catalog # PIER48895
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:50 mg
- Description:Marfey’s reagent, (1-fluoro-2-4-dinitrophenyl-5-L-alanine amide)
- Environmentally Preferable:
Specifications
About this item
Marfey's Reagent (FDAA or 1-fluoro-2-4-dinitrophenyl-5-L-alanine amide) reacts with primary amines to enable the quick and easy separation and quantitation of optical isomers of amino acids by reverse-phase chromatography.
- For precolumn derivatisation of primary amines for detection by UV absorbance (λmax = 340nm)
- Provides nanomole sensitivity in typical HPLC applications with amino acids
- Reaction products enable chiral separation of D- and L-amino acids
- Following reverse phase HPLC, derivatives have an extinction coefficient of 3×104 M-1 cm-1