1,3,5-Tris[(3,3,3-trifluoropropyl)methyl]cyclotrisiloxane 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 1,3,5-Tris(3,3,3-trifluoropropyl)-1,3,5-trimethylcyclotrisiloxane, 1,3,5-Trimethyl-1,3,5-tris(3,3,3-trifluoropropyl)cyclotrisiloxane
CAS Number:2374-14-3
:
Formula:C₁₂H₂₁F₉O₃Si₃
MDL Number:MFCD00054667
Boiling Pt:135 °C (22 mmHg)
Melting Pt:34…36 °C
Density:1.240
Molecular Weight:468.54 g/mol
Storage Temperature:Ambient
EINECS:219-154-7
ADR:6.1 III
:GHS06
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together