Tween 20 washing buffer, 1,0% in PBS (20X)
- Environmentally Preferable:
Frequently Bought Together
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
E/E 70% 70 v/v, TechniSolv®
: VWR Chemicals
VWR®, Cell Culture Flasks, Non Treated or Treated
: VWR Collection
Ethanol ≥96% (v/v), TechniSolv®, pure
: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
: VWR Collection
2-Propanol ≥98%, TECHNICAL
: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Protein coated cell culture plates, CELLCOAT®, collagen type I
: Greiner Bio-One