Anti-ABCE1 Rabbit Polyclonal Antibody
BOSSBS-2948R
: Bioss
- Antibody type:Primary
- Antigen name:ATP Binding Cassette, Subfamily E, Member 1
- Antigen symbol:ABCE1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Gene ID:6059
- Antigen synonyms:RNase 4 inhibitor|Ribonuclease 4 Inhibitor|ABC 38|ABCE 1|OABP|ATP Binding Cassette Sub Family E Member 1|RLI|RNS4I|ABC38|RNase L Inhibitor
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
ABCE1 is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the OABP subfamily. It is alternatively referred to as the RNase L inhibitor as it functions to block the activity of ribonuclease L(referenced from entrez gene).
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: ABCE1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat