Order Entry
Austria
ContactUsLinkComponent
 
Anti-CD26 Antibody
  BSBTPB9580
undefined
Anti-CD26 Antibody
  BSBTPB9580

 

  • Antibody type:
    Primary
  • Antigen name:
    Dipeptidylpeptidase 4
  • Antigen symbol:
    DPP4
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Gene ID:
    1803
  • Antigen synonyms:
    ADCP2|ADABP|DPPIV|TP103|CD26
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Rabbit IgG polyclonal antibody for Dipeptidyl peptidase 4(DPP4) detection. Tested with WB in Human;Rat.

Type: Primary
Antigen: CD26
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat