Order Entry
Austria
ContactUsLinkComponent
Anti-PPM1F Antibody Pair
Anti-PPM1F Antibody Pair
  ABNOH00009647-AP51
 :  Abnova
Anti-PPM1F Antibody Pair
  ABNOH00009647-AP51
 :  Abnova

 

  • Antigen name:
    Protein Phosphatase, Mg2+/Mn2+ dependent 1F
  • Antigen symbol:
    PPM1F
  • Host:
    Multiple
  • Isotype:
  • Reactivity:
    Human
  • Environmentally Preferable:
  • Gene ID:
    9647
  • Antigen synonyms:
    POPX2|Partner of PIX 2|hFEM-2|CaMKPase|Ca(2+)/calmodulin-dependent protein kinase phosphatase|CaM-kinase phosphatase|Protein phosphatase 1F
  • Storage temperature:
    Store reagents of the antibody pair set at –20 °C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to –20 °C storage immediately after use.
  • Size:
    1 Set
  • Pk:
    1 SET

 

 

This Set comes with matched Antibody Pair to detect and quantify protein level of human PPM1F.

Antibody pair set content: 1. Capture Antibody: Mouse Monoclonal Anti-PPM1F (100 µg); 2. Detection Antibody: Rabbit purified Polyclonal Anti-PPM1F (50 µg);

Type:
Antigen: PPM1F
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human