Order Entry
Austria
ContactUsLinkComponent
Anti-ABCC1 Goat Polyclonal Antibody
Anti-ABCC1 Goat Polyclonal Antibody
Catalog # ABNOPAB11480
Supplier:  Abnova
Anti-ABCC1 Goat Polyclonal Antibody
Catalog # ABNOPAB11480
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    ATP-binding Cassette, Subfamily C (CFTR/MRP), Member 1
  • Antigen symbol:
    ABCC1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    4363
  • Antigen synonyms:
    multiple drug resistance-associated protein|ABC29|DKFZp781G125|DKFZp686N04233|MRP1|LTC4 transporter|multidrug resistance protein|ABCC|sub-family C (CFTR/MRP)|sub-family C|member 1|GS-X|ATP-binding cassette|leukotriene C(4) transporter|multiple drug resistance protein 1|MRP|ABCC1
  • Storage buffer:
    Tris saline, pH 7,3 (0,5% BSA, 0,02% sodium azide)
  • Sequence:
    C-HQSDLKVDENQKAYY
  • Storage temperature:
    Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    A synthetic peptide corresponding to human ABCC1.
  • Purification:
    Antigen affinity purification
  • Size:
    100 μg
  • Pk:
    100 µg

Specifications

About this item

Goat polyclonal antibody raised against synthetic peptide of ABCC1.

Recommended Dilutions: ELISA: 1:128000; Western Blot: 0.05-0.2 ?g/ml

Type: Primary
Antigen: ABCC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human