Order Entry
Austria
ContactUsLinkComponent
Anti-SYNGAP Goat Polyclonal Antibody
Anti-SYNGAP Goat Polyclonal Antibody
  ABNOPAB24554
 :  Abnova
Anti-SYNGAP Goat Polyclonal Antibody
  ABNOPAB24554
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    Synaptic Ras GTPase-activating protein 1
  • Antigen symbol:
    SYNGAP
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • Isotype:
  • Reactivity:
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    8831
  • Antigen synonyms:
    ras GTPase-activating protein SynGAP|SYNGAP1|MRD5|DKFZp761G1421|synaptic Ras GTPase activating protein 1 homolog|synaptic Ras GTPase activating protein 1|KIAA1938|neuronal RasGAP|RASA5|RASA1
  • Amino acid number:
    1169 to 1183
  • Storage buffer:
    0,5 mg/mL Tris saline, pH 7,3 (0,02% sodium azide, 0,5% BSA)
  • Sequence:
    C-ESAHIEREEYKLKEY
  • Storage temperature:
    Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    A synthetic peptide corresponding to amino acids 1169-1183 near C-terminus of human SYNGAP1.
  • Purification:
    Antigen affinity purification
  • Size:
    100 μg
  • Pk:
    100 µg

 

 

Goat polyclonal antibody raised against synthetic peptide of SYNGAP1.

Recommended Dilutions: ELISA: 1:32000; Western Blot: 0.03-0.1 ?g/ml

Type: Primary
Antigen: SYNGAP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse