Anti-MDK Mouse Polyclonal Antibody
Catalog # ABNOH00004192-B01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Midgestation And Kidney Protein
- Antigen symbol:MDK
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:4192
- Antigen synonyms:ARAP|NEGF2|MK
- Amino acid number:1 to 143
- Storage buffer:1x PBS, pH 7,4
- Sequence:MQHRGFLLLTLLALLALTSAVAKKKDKVKKGGPGSECAEWAWGPCAPSSKDCGVGFREGTCGAQTQRIRCRVPCNWKKEFGADCKYKFENWGACDGGTGTKVRQGTLKKARYNAQCQETIRVTKPCTPKTKAKAKAKKGKGKD
- Storage temperature:Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:MDK (AAH11704, 1 a.a. ~ 143 a.a) full-length human protein.
- Size:50 μg
- Pk:50 µg
Specifications
About this item
Mouse polyclonal antibody raised against a full-length human MDK protein.
Recommended Dilutions: Western Blot: 1 ?g/ml
Type: Primary
Antigen: MDK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human