Order Entry
Austria
ContactUsLinkComponent
Anti-ISG15 Mouse Monoclonal Antibody [clone: 3E5]
Anti-ISG15 Mouse Monoclonal Antibody [clone: 3E5]
Catalog # ABNOMAB1090
Supplier:  Abnova
Anti-ISG15 Mouse Monoclonal Antibody [clone: 3E5]
Catalog # ABNOMAB1090
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    ISG15 Ubiquitin-like Modifier
  • Antigen symbol:
    ISG15
  • Clonality:
    Monoclonal
  • Clone:
    3E5
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    9636
  • Antigen synonyms:
    interferon-induced 17 kDa protein|ubiquitin cross-reactive protein|interferon|alpha-inducible protein (clone IFI-15K)|IP17|hUCRP|interferon-stimulated protein|UCRP|IFI15|G1P2|interferon-induced 15 kDa protein|interferon-induced 17-kDa/15-kDa protein|15 kDa|ubiquitin-like protein ISG15
  • Amino acid number:
    1 to 157
  • Storage buffer:
    PBS, pH 7,4 (0,1% sodium azide)
  • Storage temperature:
    Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Recombinant protein corresponding to amino acids 1-157 of human ISG15.
  • Size:
    100 μl
  • Pk:
    100 µg

Specifications

About this item

Mouse monoclonal antibody raised against partial recombinant ISG15.

Recommended Dilutions: Western Blot: 1:1000-1:2000

Type: Primary
Antigen: ISG15
Clonality: Monoclonal
Clone: 3E5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human