Anti-CAMK2A Monoclonal Antibody [clone: 22B1]
ABNOMAB6627
: Abnova
- Antibody type:Primary
- Antigen name:Calcium/Calmodulin-dependent Protein Kinase II alpha
- Antigen symbol:CAMK2A
- Clonality:Monoclonal
- Clone:22B1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Rat
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Antigen synonyms:calcium/calmodulin-dependent protein kinase (CaM kinase) II alpha|calcium/calmodulin-dependent protein kinase II alpha-B subunit|calcium/calmodulin-dependent protein kinase II alpha|Calci|calcium/calmodulin-dependent protein kinase type II alpha chain
- Storage buffer:PBS, pH 7,4 (50% glycerol, 0,09% sodium azide)
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to CaMKII.
- Size:100 μg
- Pk:100 µl
Mouse monoclonal antibody raised against synthetic peptide of CaMKII.
Recommended Dilutions: Western Blot: 1 ?g/ml
Type: Primary
Antigen: CAMK2A
Clonality: Monoclonal
Clone: 22B1
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1
Reactivity: Rat