Anti-ABCA1 Mouse Monoclonal Antibody [clone: AB.H10]
Catalog # ABNOMAB8772
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:ATP Binding Cassette, sub-family A, member 1
- Antigen symbol:ABCA1
- Clonality:Monoclonal
- Clone:AB.H10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:
- Reactivity:Chicken,Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-Terminal
- Cross adsorption:No
- Format:Liquid
- Gene ID:19
- Antigen synonyms:ABC-1|ATP-binding cassette transporter 1|CERP|HDLDT1|TGD|ABC1|membrane-bound|ATP-binding cassette transporter A1|cholesterol efflux regulatory protein
- Amino acid number:1800 to 2260
- Storage buffer:PBS, pH 7,4 (0,05% sodium azide)
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to amino acids 1800-2260 at C-terminus end of human ABCA1.
- Purification:Protein A purification
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Mouse monoclonal antibody raised against synthetic peptide of ABCA1.
Recommended Dilutions: Western Blot: 1:1000
Type: Primary
Antigen: ABCA1
Clonality: Monoclonal
Clone: AB.H10
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Mouse
Isotype:
Reactivity: Chicken, Human, Mouse