Anti-RAD51AP1 Goat Polyclonal Antibody
Catalog # ABNOPAB6275
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:RAD51 Associated Protein 1
- Antigen symbol:RAD51AP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:10635
- Antigen synonyms:PIR51
- Storage buffer:Tris saline, pH 7,3 (0,5% BSA, 0,02% sodium azide)
- Sequence:C-ARVKPLHPNATST
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to C-terminus of human RAD51AP1.
- Purification:Antigen affinity purification
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Goat polyclonal antibody raised against synthetic peptide of RAD51AP1.
Recommended Dilutions: ELISA: 1:32000; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 4-6 ?g/ml
Type: Primary
Antigen: RAD51AP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human