Anti-ASAP2 Rabbit Polyclonal Antibody
ABNOPAB15764
: Abnova
- Antibody type:Primary
- Antigen name:ArfGAP With SH3 Domain, Ankyrin Repeat And PH Domain 2
- Antigen symbol:ASAP2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:211914
- Antigen synonyms:Pap-alpha|SHAG1|PAP|AMAP2|CENTB3|DDEF2|PAG3
- Storage buffer:PBS (50% glycerol, 0,02% sodium azide)
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant GST fusion protein corresponding to 251 amino acids of mouse Asap2.
- Size:50 μg
- Pk:50 µg
Rabbit polyclonal antibody raised against partial recombinant Asap2.
Recommended Dilutions: Western Blot: 1:1000
Type: Primary
Antigen: ASAP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse