Anti-TMEM59 Rabbit Polyclonal Antibody
Catalog # ABNOPAB19345
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Transmembrane Protein 59
- Antigen symbol:TMEM59
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:9528
- Antigen synonyms:Transmembrane protein 59|C1orf8|HSPC001|Liver membrane-bound protein|UNQ169/PRO195|TMEM59
- Storage buffer:PBS (0,02% sodium azide)
- Storage temperature:Store at 4 °C for three months. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to 16 amino acids near N-terminus of human TMEM59.
- Purification:Peptide affinity purification
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Rabbit polyclonal antibody rasied against synthetic peptide of TMEM59.
Recommended Dilutions: Western Blot: 1 ?g/ml; Immunohistochemistry: 2.5 ?g/ml
Type: Primary
Antigen: TMEM59
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human