Anti-TACR1 Goat Polyclonal Antibody
Catalog # ABNOPAB7270
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Tachykinin Receptor 1
- Antigen symbol:TACR1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:6869
- Antigen synonyms:TACR1|TAC1R |NK1R|NKIR|neurokinin 1 receptor|NK-1 receptor|tachykinin 1 receptor (substance P receptor|SPR|tachykinin receptor 1|Tachykinin receptor 1 (substance P receptor,neurokinin-1 receptor)|neurokinin 1 receptor)
- Storage buffer:Tris saline, pH 7,3 (0,5% BSA, 0,02% sodium azide)
- Sequence:C-EIPGDSSDRYHEQ
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to human TACR1.
- Purification:Antigen affinity purification
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Goat polyclonal antibody raised against synthetic peptide of TACR1.
Recommended Dilutions: ELISA: 1:32000; Western Blot: 1-3 ?g/ml
Type: Primary
Antigen: TACR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human