Anti-PLCG2 Rabbit Polyclonal Antibody
ABNOPAB25823
: Abnova
- Antibody type:Primary
- Antigen name:Phospholipase C Gamma 2
- Antigen symbol:PLCG2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:5336
- Antigen synonyms:PLC-IV|Phospholipase C-IV|PLCG2|Phospholipase C-gamma-2|PLC gamma-2|Phosphoinositide phospholipase C-gamma-2|PLC-gamma-2|1-phosphatidylinositol 4,5-bisphosphate phosphodiesterase gamma-2|PLC gamma2|PLC- gamma2
- Modification:phospho-Tyr753
- Storage buffer:PBS (without Mg(2+) and Ca(2+)), 150 mM NaCl, pH 7,4 (50% glycerol, 0,02% sodium azide)
- Sequence:S-L-Yp-D-V
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Synthetic phosphopeptide corresponding to residues surrounding Y753 of human PLCG2.
- Purification:purified by Affinity Chromatography
- Size:100 μg
- Pk:100 µg
Rabbit polyclonal antibody raised against synthetic phosphopeptide of PLCG2.
Recommended Dilutions: Western Blot: 1:500-1:1000
Type: Primary
Antigen: PLCG2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat