Order Entry
Austria
ContactUsLinkComponent
Anti-CD38 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: HIT2]
Anti-CD38 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: HIT2]
Supplier:  Tonbo Biosciences
Certificates
Anti-CD38 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: HIT2]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 50-0389-T025
  • 50-0389-T100
  • Antigen symbol:
  • CD38
  • CD38
  • Antigen name:
  • CD38 molecule
  • CD38 molecule
  • Antigen synonyms:
  • NAD+ nucleosidase|ADP-ribosyl cyclase 1|T10|cADPr hydrolase 1|ADP-ribosyl cyclase/cyclic ADP-ribose hydrolase|cyclic ADP-ribose hydrolase 1|CD38 antigen p45
  • NAD+ nucleosidase|ADP-ribosyl cyclase 1|T10|cADPr hydrolase 1|ADP-ribosyl cyclase/cyclic ADP-ribose hydrolase|cyclic ADP-ribose hydrolase 1|CD38 antigen p45
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • PE (Phycoerythrin)
  • PE (Phycoerythrin)
  • Clone:
  • HIT2
  • HIT2
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 952
  • 952
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Flow cytometry:
  • Yes
  • Yes
  • Storage temperature:
  • +2 °C to +8 °C, protected from light
  • +2 °C to +8 °C, protected from light

Specifications

Related Information

Frequently Bought Together