The 17A2 antibody reacts with the mouse CD3 complex, comprised of CD3 epsilon, CD3 gamma and CD3 delta. These integral membrane protein chains assemble with additional chains of the T cell receptor (TCR), as well as CD3 zeta chain, to form the T cell receptor – CD3 complex. Together with co-receptors CD4 or CD8, the complex serves to recognize antigens bound to MHC molecules on antigen-presenting cells. Such interactions promote T cell receptor signaling (T cell activation) and can result in a number of cellular responses including proliferation, differentiation, production of cytokines or activation-induced cell death. CD3 is differentially expressed during thymocyte-to-T cell development and on all mature T cells.
The 17A2 antibody is a widely used phenotypic marker for mouse T cells. In addition, as the CD3e chain within the TCR complex contains intracellular signaling domains, binding of 17A2 antibody to CD3 can induce cell activation (use format suitable for functional assays). A recent publication of the crystal structure of a murine CD3e-mitogenic antibody complex provides further insight into the action of commonly used agonist antibodies (Fernandes, R.A. et al. 2012. Journal of Biological Chemistry. 287: 13324-13335).
Type:
Antigen: CD3
Clonality:
Clone: 17A2
Conjugation: redFluor™ 710
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
- Catalog No:
- 80-0032-U025
- 80-0032-U100
- Antigen symbol:
- CD3G
- CD3G
- Antigen name:
- CD3 antigen, gamma polypeptide
- CD3 antigen, gamma polypeptide
- Antigen synonyms:
- T3g|Ctg3|Ctg-3
- T3g|Ctg3|Ctg-3
- Conjugation:
- redFluor® 710
- redFluor® 710
- Clone:
- 17A2
- 17A2
- Reactivity:
- Mouse
- Mouse
- Host:
- Rat
- Rat
- Gene ID:
- 12502
- 12502
- Isotype:
- IgG2b
- IgG2b
- Flow cytometry:
- Yes
- Yes
- Storage temperature:
- Store Undiluted at 2 to 8 °C
- Store Undiluted at 2 to 8 °C