Anti-GAPDH Mouse Monoclonal Antibody [clone: 1A10A10]
ABGEAM1020B
: Abgent
- Antibody type:Primary
- Antigen name:Glyceraldehyde-3-phosphate dehydrogenase
- Antigen symbol:GAPDH
- Clonality:Monoclonal
- Clone:1A10A10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Gene ID:2597
- Antigen synonyms:G3PD|GAPD|HEL-S-162eP
- Storage buffer:pH 7,4
- Molecular weight:36.053 kDa
- Sequence:Unknown
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0.44
- Shipping temperature:4 °C
- Immunogen:GAPDH recombinant protein is used to produce this monoclonal antibody.
- Purification:Protein G Purification
- Pk:400 µl
Type: Primary
Antigen: GAPDH
Clonality: Monoclonal
Clone: 1A10A10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat