Anti-AKT3 Mouse Monoclonal Antibody (Unconjugated) [clone: 25F6.F6.D8]
Catalog # ROCK200-301-E75S
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:v-akt murine thymoma viral oncogene homolog 3
- Antigen symbol:AKT3
- Clonality:Monoclonal
- Clone:25F6.F6.D8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Gene ID:10000
- Antigen synonyms:PKB-GAMMA|PKBG|RAC-PK-gamma|STK-2|RAC-gamma|PRKBG|MPPH2|MPPH
- Size:25 µl
- Pk:25 µl
Specifications
About this item
Type: Primary
Antigen: AKT3
Clonality: Monoclonal
Clone: 25F6.F6.D8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human