Anti-FITC Goat Polyclonal Antibody (DyLight® 488)
ROCK600-141-096
- Antibody type:Tag
- Antigen name:Fluorescein
- Antigen symbol:FITC
- Clonality:Polyclonal
- Conjugation:DyLight® 488
- Host:Goat
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Cat
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Antibody fraction
- Form:Lyophilized
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin, Immunoglobulin and Protease free
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:Fluorescein conjugated to Goat IgG
- Tested applications:Fluoresence ELISA
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Anti-Fluorescein Antibody Dylight® 488 Conjugated is designed for immunofluorescence microscopy, fluorescence based plate assays (FLISA) and fluorescent western blotting. This product is also suitable for multiplex analysis, including multicolor imaging.
Recommended Dilutions: FLISA: >1:20000; Immunofluorescence: >1:5000; Western Blot: 1:10000 - 1:25000;
Restore with deionized water (or equivalent); 100 µl
Type: Tag
Antigen: FITC
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Goat
Isotype: IgG
Reactivity: