Anti-Filaggrin 2 Rabbit Polyclonal Antibody
Catalog # USBI140329
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Filaggrin 2
- Antigen symbol:FLG2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Amino acid number:1915 to 2143
- Storage buffer:PBS, pH 7,2, 0,05% sodium azide, 50% glycerol.
- Sequence:MGHHHHHHSGSEFVHKRHQTTHGQTGDTTEHGHPSHGQTIQTGSRTTGRRGSGHSEYSDSEGPSGVSHTHSGHTHGQAGSHYPESGSSVHERHGTTHGQTADTTRHGHSGHGQSTQRGSRTTGRRASGHSEYSDSEGHSGVSHTHSGHAHGQAGSQHGESGSSVHERHGTTHGQTGDTTRHAHSGHGQSTQRGSRTAGRRGSGHSESSDSEVHSGVSHTHSGHTYGQARSQHGESGSAIHGR
- Storage temperature:–20 °C
- Concentration:200 ug/ml
- Immunogen:FLG2 (Val1915~Arg2143); MGHHHHHHSGSEF-VHKRHQ TTHGQTGDTT EHGHPSHGQT IQTGSRTTGR RGSGHSEYSD SEGPSGVSHT HSGHTHGQAG SHYPESGSSV HERHGTTHGQ TADTTRHGHS GHGQSTQRGS RTTGRRASGH SEYSDSEGHS GVSHTHSGHA HGQAGSQHGE SGSSVHERHG TTHGQTGDTT RHAHSGHGQS TQRGSRTAGR RGSGHSESSD SEVHSGVSHT HSGHTYGQAR SQHGESGSAI HGR
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Recommended dilution: ELISA: 1:100-200; Western Blot: 1:100-400; Immunochemistry: 1:100-500
Type: Primary
Antigen: Filaggrin 2
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human,