Order Entry
Austria
ContactUsLinkComponent
Anti-Heparan-Sulfate Proteoglycan Mouse Monoclonal Antibody [clone: 2H7/G11]
Anti-Heparan-Sulfate Proteoglycan Mouse Monoclonal Antibody [clone: 2H7/G11]
  USBIH1890-93B-100
Anti-Heparan-Sulfate Proteoglycan Mouse Monoclonal Antibody [clone: 2H7/G11]
  USBIH1890-93B-100

 

  • Antibody type:
    Primary
  • Antigen name:
    Heparan-Sulfate Proteoglycan
  • Clonality:
    Monoclonal
  • Clone:
    2H7/G11
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Form:
    lyophilized powder
  • Storage buffer:
    50mMTBS, pH 7,2.
  • Storage temperature:
    –20 °C
  • Concentration:
    ~1 mg/ml
  • Immunogen:
    Purified human small basement membrane heparan sulfate proteoglycan (HS-PG)
  • Purification:
    purified by Protein G chromatography
  • Size:
    100 μg
  • Pk:
    100 µG

 

 

Recommended dilution: ELISA: 1 ?g/ml; Western Blot: 1 ?g/ml; Immunochemistry: 1 ?g/ml

Type: Primary
Antigen: Heparan-Sulfate Proteoglycan
Clonality: Monoclonal
Clone: 2H7/G11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human,