Test tube racks, Switch-Grid, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
Certificates
About this item
Polyoxymethylene, coloured.
- Test tubes with 10 to 30 mm diameter fit in 5 colour coded grid inserts
- Three tier rack holds tubes securely in place
- Rubber feet keep racks from sliding on benchtop and also provide secure stacking while in storage
- Can be cleaned with mild detergent, ethanol or isopropyl alcohol
The innovative, split-frame rack holds up to two different size tubes in the same rack and allows users to quickly switch grids to hold other sizes as needed. Chemical resistant polyoxymethylene plastic rack frame and grids can be cleaned with mild detergent, ethanol or isopropyl alcohol.
Ordering information: Each rack frame includes a set of grids in one size. Grid sets in alternate sizes must be ordered separately.
Frequently Bought Together