ContactUsLinkComponent
Flow meters, soap bubble
Flow meters, soap bubble
Catalog # PERC00230522
Supplier:  PerkinElmer
Flow meters, soap bubble
Catalog # PERC00230522
Supplier:  PerkinElmer
Specifications
  • Description:
    Soap bubble flow meter (two stage) 1 and 10 ml
  • Seq:
    02
  • Environmentally Preferable:
  • Pk:
    1
Specifications
About this item
Two stage soap bubble flow meter.
Delivery: Supplied with liquid soap, rubber squeeze bulb, plastic tubes and instruction manual.