Anti-Acetyl Lysine Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: 7F8]
COBSD5-1762
- Antibody type:Primary
- Antigen name:Acetyl-Lysine
- Antigen symbol:AcK
- Clonality:Monoclonal
- Clone:7F8
- Conjugation:RPE (R-Phycoerythrin)
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:
- Environmentally Preferable:
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: Acetyl Lysine
Clonality: monoclonal
Clone: 7F8
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: