Anti-Solute Carrier Family 23A2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # USBIS5329-23D-HRP
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:Solute Carrier Family 23A2
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-terminal
- Form:liquid
- Storage buffer:PBS, pH 7,2
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500
Type: Primary
Antigen: Solute Carrier Family 23A2
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human