Anti-POU Domain, Class 4, Transcription Factor 1 Mouse Monoclonal Antibody [clone: 9G417]
Catalog # USBIP5250-30A
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:POU Domain, Class 4, Transcription Factor 1
- Clonality:Monoclonal
- Clone:9G417
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:lyophilized powder
- Storage buffer:PBS, pH 7,2
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Immunochemistry: 10 ?g/ml
Type: Primary
Antigen: POU Domain, Class 4, Transcription Factor 1
Clonality: monoclonal
Clone: 9G417
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human