Order Entry
Austria
ContactUsLinkComponent
Anti-Phosphoinositide 3 Kinase, p110 alpha Rabbit Polyclonal Antibody
Anti-Phosphoinositide 3 Kinase, p110 alpha Rabbit Polyclonal Antibody
Catalog # USBIP4073-26H
Supplier:  US Biological
Anti-Phosphoinositide 3 Kinase, p110 alpha Rabbit Polyclonal Antibody
Catalog # USBIP4073-26H
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Phosphoinositide 3 Kinase, p110 alpha
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Form:
    liquid
  • Storage buffer:
    10mMsodium HEPES, pH 7,5, 150mM sodium chloride, 0,1mg/ml BSA, 50% glycerol.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A chromatography
  • Size:
    100 µl
  • Pk:
    100 µl

Specifications

About this item

Recommended dilution: Western Blot: 1:1000; Immunoprecipitation: 1:100

Type: Primary
Antigen: Phosphoinositide 3 Kinase, p110 alpha
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat