Order Entry
Austria
ContactUsLinkComponent
Anti-NGFI-A Binding Protein 1 Mouse Monoclonal Antibody [clone: 8C238]
Anti-NGFI-A Binding Protein 1 Mouse Monoclonal Antibody [clone: 8C238]
Catalog # USBIN2414-10
Supplier:  US Biological
Anti-NGFI-A Binding Protein 1 Mouse Monoclonal Antibody [clone: 8C238]
Catalog # USBIN2414-10
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    NGFI-A Binding Protein 1
  • Clonality:
    Monoclonal
  • Clone:
    8C238
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human,
    Rat
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,4, 1% BSA, 0,05% sodium azide.
  • Storage temperature:
    Cool
  • Concentration:
    ~0,1 mg/ml
  • Shipping temperature:
    Ice
  • Tested applications:
    Dot Blot
  • Purification:
    purified by Protein G chromatography
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

Recommended dilution: Western Blot: 0.2-2?g/ml; Immunochemistry: 2-100 ?g/ml

Type: Primary
Antigen: NGFI-A Binding Protein 1
Clonality: monoclonal
Clone: 8C238
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat