Anti-NEI-MCH Rabbit Polyclonal Antibody
USBIN0590-32A
- Antibody type:Primary
- Antigen symbol:NEI-MCH
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Form:lyophilized powder
- Amino acid number:131 to 165
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified
- Size:200 µg
- Pk:200 µG
Type: Primary
Antigen: NEI-MCH
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat