Order Entry
Austria
ContactUsLinkComponent
 
Anti-Matrix Metalloproteinase 1 Mouse Monoclonal Antibody
  USBIM2420-05B
undefined
Anti-Matrix Metalloproteinase 1 Mouse Monoclonal Antibody
  USBIM2420-05B

 

  • Antibody type:
    Primary
  • Antigen name:
    Matrix Metalloproteinase 1
  • Antigen symbol:
    MMP1
  • Clonality:
    Monoclonal
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,2, 0,09% sodium azide.
  • Storage temperature:
    Cool
  • Concentration:
    ~0,5 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Recommended dilution: Western Blot: 4?g/ml

Type: Primary
Antigen: Matrix Metalloproteinase 1
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human