Order Entry
Austria
ContactUsLinkComponent
Anti-MAP Kinase 14 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Anti-MAP Kinase 14 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
  USBIM2352-03U-PE
undefined
Anti-MAP Kinase 14 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
  USBIM2352-03U-PE

 

  • Antibody type:
    Primary
  • Antigen name:
    Mitogen-activated protein kinase 14
  • Antigen symbol:
    MAPK14
  • Clonality:
    Polyclonal
  • Conjugation:
    PE (Phycoerythrin)
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,2
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Tested applications:
    Fluoresence ELISA
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    200 µl
  • Pk:
    200 µl

 

 

Recommended dilution: FLISA: 1:1000; Western Blot: 1:50-1:100
Do not freeze!

Type: Primary
Antigen: MAP Kinase 14
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human