Anti-Golgi Associated, Gamma Adaptin Ear Containing, ARF Binding Protein 3 Goat polyclonal antibody
USBIG8576-88C
- Antibody type:Primary
- Antigen name:Golgi Associated, Gamma Adaptin Ear Containing, ARF Binding Protein 3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:lyophilized powder
- Amino acid number:489 to 656
- Storage buffer:0,2um sterile-filtered solution
- Storage temperature:Cool
- Concentration:~0,2 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Recommended dilution: ELISA: 0.5-1.0 ?g/ml; Western Blot: 1.0 ?g/ml
Type: Primary
Antigen: Golgi Associated, Gamma Adaptin Ear Containing, ARF Binding Protein 3
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human