Anti-Bradykinin Receptor Type B1 Goat polyclonal antibody unconjugated
USBIB2675-80B
- Antibody type:Primary
- Antigen name:Bradykinin Receptor Type B1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Storage buffer:Tris saline, pH 7,2, 0,5% BSA, 0,02% sodium azide.
- Sequence:KVWELYKQCTPK
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Recommended dilution: ELISA: 1:1000; Western Blot: 2-4?g/ml
Type: Primary
Antigen: Bradykinin Receptor Type B1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human