Anti-PTBP3 Mouse Monoclonal Antibody [clone: 4C9]
USBI132722
- Antibody type:Primary
- Antigen name:Polypyrimidine tract binding protein 3
- Antigen symbol:PTBP3
- Clonality:Monoclonal
- Clone:4C9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Amino acid number:16 to 114
- Storage buffer:PBS, pH 7,2.
- Sequence:GSDELLSSGIINGPFTMNSSTPSTANGNDSKKFKRDRPPCSPSRVLHLRKIPCDVTEAEIISLGLPFGKVTNLLMLKGKSQAFLEMASEEAAVTMVNYY
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Recommended dilution: Immunofluorescence: 20 ?g/ml; Immunochemistry: 3?g/ml
Type: Primary
Antigen: PTBP3 (polypyrimidine tract binding protein 3)
Clonality: monoclonal
Clone: 4C9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse, Rat