Anti-GNL2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
USBI036145-FITC
- Antibody type:Primary
- Antigen name:Guanine nucleotide binding protein-like 2 (nucleolar)
- Antigen symbol:GNL2
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-terminal
- Form:liquid
- Amino acid number:17 to 45
- Storage buffer:PBS, pH 7,2, contains ~0,09% sodium azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:200 µl
- Pk:200 µl
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: GNL2 (guanine nucleotide binding protein-like 2 (nucleolar))
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human