Anti-GCNT2 Rabbit Polyclonal Antibody
Catalog # USBI035967
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Glucosaminyl (N-acetyl) transferase 2, I-branching enzyme (I blood group)
- Antigen symbol:GCNT2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-terminal
- Form:liquid
- Amino acid number:371 to 399
- Storage buffer:PBS, pH 7,2, 0,09% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100
Type: Primary
Antigen: GCNT2 (glucosaminyl (N-acetyl) transferase 2, I-branching enzyme (I blood group))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse