Anti-DTX3L Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # USBI034842-HRP
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Deltex 3 like, E3 ubiquitin ligase
- Antigen symbol:DTX3L
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:internal
- Form:liquid
- Amino acid number:452 to 482
- Storage buffer:PBS, pH 7,2
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Type: Primary
Antigen: DTX3L (deltex 3 like, E3 ubiquitin ligase)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human