Anti-Canine Serum Albumin Rabbit Polyclonal Antibody
Catalog # USBI028979
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Canine Serum Albumin (Canine Serum Albumin)
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Dog
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Storage buffer:PBS.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by ammonium sulfate precipitation
- Size:50 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: Canine Serum Albumin (Canine Serum Albumin)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Dog