Anti-CD69 Mouse Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: FN50]
Catalog # ROCK200-302-N73
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:CD69 molecule
- Antigen symbol:CD69
- Clonality:Monoclonal
- Clone:FN50
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Environmentally Preferable:
- Cross adsorption:none
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Gene ID:969
- Antigen synonyms:EA1|MLR-3|AIM|GP32/28|CLEC2C|BL-AC/P26
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,09% (w/v) Sodium Azide, 0,2% BSA (w/v)
- Storage temperature:Cool
- Concentration:Sufficient to run approximately 100 tests
- Shipping temperature:Ice
- Immunogen:Anti-CD69 Antibody (Monoclonal) was produced by repeated immunizations with CD69 antigen.
- Tested applications:Fluoresence ELISA
- Purification:affinity-purified
- Size:500 µl
- Pk:500 µl
Specifications
About this item
Anti-CD69 is useful for Flow Cytometry, immunochemistry, and Immunofluorescence. Researchers should determine optimal titers for applications that are not stated.
Recommended Dilutions: Flow Cytometry: 5 µl/test/1x10e5 to 1x10e8 cells;
Do not freeze!
Type: Primary
Antigen: CD69
Clonality: Monoclonal
Clone: FN50
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human